Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011646.5 | NP_035776.1 | 5 | 774 | ||
GenBank mRNA | AK003064.1 | - | 5 | 778 | ||
AK008660.1 | - | 5 | 775 | |||
AK008667.1 | - | 5 | 775 | |||
BC050120.1 | - | 5 | 755 | |||
BC061135.1 | - | 5 | 774 | 30306436 |