Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011653.2 | NP_035783.1 | 4 | 1400 | ||
GenBank mRNA | AK077529.1 | - | 4 | 1412 | ||
AK145404.1 | - | 4 | 1449 | |||
AK146983.1 | - | 4 | 1449 | |||
AK161783.1 | - | 2 | 996 | |||
AK164335.1 | - | 4 | 1451 | |||
AK167687.1 | - | 4 | 1449 | |||
AK168762.1 | - | 4 | 1449 | |||
AK169610.1 | - | 4 | 1450 | |||
AK216974.1 | - | 1 | 196 | |||
BC056169.1 | - | 4 | 1411 | 5715646 | ||
BC083343.1 | - | 4 | 1424 | 5687414 | ||
BC083344.1 | - | 4 | 1419 | 5693701 | ||
BC083345.1 | - | 4 | 1434 | 6415067 | ||
BC085256.1 | - | 4 | 1433 | 6306481 | ||
M13445.1 | - | 4 | 1400 | |||
M28729.1 | - | 1 | 593 |