Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009448.4 | NP_033474.1 | 4 | 1875 | ||
GenBank mRNA | AK013029.1 | - | 2 | 1031 | ||
AK145567.1 | - | 4 | 1475 | |||
AK167654.1 | - | 4 | 1445 | |||
AK167759.1 | - | 4 | 1408 | |||
AK191962.1 | - | 1 | 253 | |||
AK202573.1 | - | 1 | 272 | |||
M13441.1 | - | 4 | 1387 |