Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011664.3 | NP_035794.1 | 2 | 1037 | ||
GenBank mRNA | AK003190.1 | - | 3 | 810 | ||
AK007273.1 | - | 2 | 1069 | |||
AK010413.1 | - | 2 | 1039 | |||
AK010625.1 | - | 2 | 1038 | |||
AK012443.1 | - | 2 | 1039 | |||
AK013027.1 | - | 2 | 1038 | |||
AK013873.1 | - | 2 | 1036 | |||
AK132747.1 | - | 2 | 1038 | |||
AK160683.1 | - | 3 | 576 | |||
AK165313.1 | - | 2 | 1037 | |||
AK209901.1 | - | 1 | 45 | |||
BC019850.1 | - | 2 | 1006 | 5123849 | ||
BC066197.1 | - | 2 | 1067 | 6435236 | ||
BC100341.1 | - | 2 | 992 | 30787432 | ||
M81745.1 | - | 1 | 249 | |||
X51703.1 | - | 2 | 1043 |
Gene Symbol: Gm1821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088453.1 | - | 2 | 583 |