Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251987.1 | NP_001238916.1 | 18, 19 | 1578 | ||
NM_006073.3 | NP_006064.2 | 18, 19 | 1575 | |||
GenBank mRNA | AJ489257.2 | - | 18, 19 | 1473 | ||
CR749591.1 | - | 8, 9 | 334 | |||
U18985.1 | - | 18, 19 | 1279 |