Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146057.3 | NP_666169.1 | 4 | 1413 | ||
GenBank mRNA | AK152216.1 | - | 4 | 1380 | ||
AK152884.1 | - | 4 | 1390 | |||
AK167579.1 | - | 4 | 1337 | |||
AK167688.1 | - | 4 | 1276 | |||
AK169162.1 | - | 4 | 1345 | |||
AK171259.1 | - | 4 | 1405 | |||
AK219071.1 | - | 1 | 178 | |||
BC010828.1 | - | 4 | 1346 | 4220180 | ||
BC024876.1 | - | 4 | 1374 | 3155209 | ||
BC057669.1 | - | 4 | 1336 | 2655176 |