Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199573.1 | NP_001186502.1 | 8 | 1569 | ||
NM_006074.4 | NP_006065.2 | 8 | 1581 | |||
GenBank mRNA | AK290434.1 | - | 8 | 1311 | ||
AK298715.1 | - | 9 | 1261 | |||
AK298934.1 | - | 8 | 1135 | |||
AK301192.1 | - | 7 | 1052 | |||
AM040752.1 | - | 5 | 529 | |||
BC022281.1 | - | 8 | 1388 | 4718024 | ||
BC035582.1 | - | 8 | 1354 | 5583800 | ||
HQ842635.1 | - | 7 | 1202 | |||
X82200.1 | - | 8 | 1312 |