Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D17Wsu92e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033279.3 | NP_001028451.2 | 1 | 287 | ||
NM_001044719.2 | NP_001038184.1 | 1 | 287 | |||
NM_001271511.1 | NP_001258440.1 | 1 | 287 | |||
GenBank mRNA | AK143016.1 | - | 1 | 200 | ||
AK161603.1 | - | 1 | 288 | |||
AK165011.1 | - | 1 | 286 | |||
BC132324.1 | - | 2 | 213 | 40130601 | ||
BC132326.1 | - | 2 | 213 | 40130603 | ||
BC137573.1 | - | 1 | 287 | 8860592 | ||
BC137576.1 | - | 1 | 287 | 8860595 | ||
BC145403.1 | - | 1 | 287 | 9053961 | ||
BY755073.1 | - | 1 | 213 | |||
CX220817.1 | - | 1 | 62 |