Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019112.3 | NP_061985.2 | 32, 33 | 4655 | ||
XM_006722616.1 | XP_006722679.1 | 32, 33 | 4642 | |||
XM_006722617.2 | XP_006722680.1 | 32, 33 | 4642 | |||
XM_006722618.2 | XP_006722681.1 | 17, 18 | 2192 | |||
XM_011527628.2 | XP_011525930.1 | 32, 33 | 4516 | |||
XM_011527629.1 | XP_011525931.1 | 32, 33 | 4615 | |||
XM_011527630.1 | XP_011525932.1 | 32, 33 | 4642 | |||
XM_011527631.1 | XP_011525933.1 | 32, 33 | 4642 | |||
XM_011527632.1 | XP_011525934.1 | 28, 29 | 4094 | |||
XM_011527633.2 | XP_011525935.1 | 32, 33 | 4642 | |||
XM_011527636.2 | XP_011525938.1 | 17, 18 | 2190 | |||
XM_017026142.1 | XP_016881631.1 | 31, 32 | 4387 | |||
GenBank mRNA | AB055390.1 | - | 26, 27 | 4010 | ||
AF250238.1 | - | 31, 32 | 4424 | |||
AF328787.1 | - | 31, 32 | 4540 |