Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019112.3 | NP_061985.2 | 43 | 6014 | ||
XM_006722616.1 | XP_006722679.1 | 43 | 5947 | |||
XM_006722618.2 | XP_006722681.1 | 28 | 3551 | |||
XM_011527628.2 | XP_011525930.1 | 43 | 5875 | |||
XM_011527629.1 | XP_011525931.1 | 43 | 5974 | |||
XM_011527630.1 | XP_011525932.1 | 42 | 5872 | |||
XM_011527631.1 | XP_011525933.1 | 40 | 5554 | |||
XM_011527632.1 | XP_011525934.1 | 39 | 5453 | |||
XM_011527636.2 | XP_011525938.1 | 28 | 3549 | |||
XM_017026142.1 | XP_016881631.1 | 42 | 5746 | |||
GenBank mRNA | AB055390.1 | - | 37 | 5369 | ||
AF250238.1 | - | 42 | 5783 | |||
AF328787.1 | - | 42 | 5899 | |||
AK097344.1 | - | 10 | 1077 | |||
AL137481.1 | - | 6 | 628 |