Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011731.3 | NP_035861.2 | 5 | 947 | ||
GenBank mRNA | AF075261.1 | - | 5 | 771 | ||
AK076806.1 | - | 6 | 681 | |||
AK143893.1 | - | 5 | 920 | |||
AK144038.1 | - | 5 | 948 | |||
AK163340.1 | - | 6 | 836 | |||
AK164706.1 | - | 5 | 792 | |||
AK164713.1 | - | 5 | 782 | |||
BC013484.1 | - | 5 | 858 | 4242168 |