Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081213.1 | NP_001074682.1 | 2 | 606 | ||
GenBank mRNA | AK037287.1 | - | 2 | 565 | ||
AK041066.1 | - | 2 | 577 | |||
AK050284.1 | - | 2 | 602 | |||
AK081674.1 | - | 1 | 123 | |||
AK129447.1 | - | 2 | 534 | |||
AK137206.1 | - | 2 | 562 | |||
BC086627.1 | - | 2 | 547 | |||
BC151181.1 | - | 2 | 565 | 9088070 | ||
BC151182.1 | - | 2 | 565 | 9088071 | ||
BC157914.1 | - | 2 | 565 | 9088152 |