Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011767.2 | NP_035897.2 | 4 | 612 | ||
GenBank mRNA | AF071059.1 | - | 4 | 610 | ||
BC038599.1 | - | 4 | 616 | |||
BC058570.1 | - | 4 | 612 | |||
BC094220.1 | - | 3 | 500 | |||
BC141139.1 | - | 4 | 596 | 9055751 |