Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144500.3 | NP_653083.2 | 7 | 705 | ||
GenBank mRNA | AK048537.1 | - | 7 | 625 | ||
AK145292.1 | - | 7 | 737 | |||
AK161695.1 | - | 7 | 644 | |||
AK166059.1 | - | 7 | 684 | |||
AK170306.1 | - | 7 | 689 | |||
AK170669.1 | - | 7 | 706 | |||
AK185626.1 | - | 2 | 95 | |||
AK220257.1 | - | 1 | 24 | |||
BC026804.1 | - | 7 | 712 | 4159217 |