Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp10d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153389.3 | NP_700438.3 | 10 | 1670 | ||
GenBank mRNA | AJ441079.1 | - | 10 | 1670 | ||
AK036646.1 | - | 9 | 1335 | |||
BC141328.1 | - | 10 | 1521 | 9055940 |