Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146191.3 | NP_666303.3 | 28 | 4653 | ||
GenBank mRNA | AK040153.1 | - | 7 | 1158 | ||
AK129441.1 | - | 25 | 4066 | |||
AK147469.1 | - | 28 | 4639 | |||
AK154395.1 | - | 2 | 172 |