Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144929.2 | NP_659178.1 | 6 | 1808 | ||
GenBank mRNA | AK080715.1 | - | 6 | 1126 | ||
AY527275.1 | - | 6 | 1808 | |||
AY527276.1 | - | 7 | 1263 | |||
BC022744.1 | - | 5 | 1380 | 4527012 | ||
BC023051.1 | - | 5 | 1336 | 5361063 | ||
BC027079.1 | - | 5 | 1336 | 5357069 |