Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172768.1 | NP_766356.1 | 19 | 2613 | ||
GenBank mRNA | AK028277.1 | - | 20 | 2508 | ||
AK033156.1 | - | 19 | 2613 | |||
AK122465.1 | - | 19 | 2585 | |||
BC030911.1 | - | 10 | 1357 | |||
BC070451.1 | - | 19 | 2459 | 30616509 |