Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AF067061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199060.1 | NP_951015.1 | 339 | |||
GenBank mRNA | AF067061.1 | - | 338 | |||
AF067062.1 | - | 339 | ||||
BC115951.1 | - | 1 | 785 | 40049464 |
Gene Symbol: Tcstv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153523.3 | NP_705743.1 | 2 | 357 | ||
GenBank mRNA | AF067060.1 | - | 355 | |||
BC125501.1 | - | 2 | 340 | 40130016 | ||
BC125503.1 | - | 2 | 340 | 40130018 |