Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011790.4 | NP_035920.1 | 14 | 1582 | ||
GenBank mRNA | AF124664.1 | - | 13 | 1409 | ||
AJ130975.1 | - | 13 | 1441 | |||
AK011094.1 | - | 14 | 1589 | |||
AK045012.1 | - | 1565 | ||||
AK149936.1 | - | 14 | 1562 | |||
AK167104.1 | - | 14 | 1539 | |||
AK168055.1 | - | 14 | 1585 | |||
AK220131.1 | - | 2 | 119 | |||
BC051998.1 | - | 14 | 1582 | 6400287 | ||
BC052422.1 | - | 14 | 1582 | 6400287 |