Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025779.1 | NP_001020950.1 | 8, 9 | 1304 | ||
NM_011799.2 | NP_035929.1 | 8, 9 | 1380 | |||
GenBank mRNA | AJ009559.1 | - | 8, 9 | 1397 | ||
AJ223087.1 | - | 8, 9 | 1382 | |||
AK012192.1 | - | 8, 9 | 1334 | |||
AK075975.1 | - | 8, 9 | 1331 | |||
AK085020.1 | - | 8, 9 | 1334 | |||
AK162669.1 | - | 8, 9 | 1342 | |||
AK166003.1 | - | 8, 9 | 1385 | |||
BC052434.1 | - | 8, 9 | 1304 | 6837113 |