Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011814.2 | NP_035944.2 | 17 | 2696 | ||
GenBank mRNA | AB025269.1 | - | 16 | 2259 | ||
AK154243.1 | - | 17 | 2695 | |||
AK192714.1 | - | 1 | 36 | |||
AK192912.1 | - | 1 | 36 | |||
AK195253.1 | - | 1 | 36 | |||
AK195317.1 | - | 1 | 36 | |||
BC057007.1 | - | 17 | 2591 | 6816943 | ||
BC062971.1 | - | 17 | 2696 | 6415619 |