Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204340.1 | NP_001191269.1 | 5, 6 | 660 | ||
NM_172810.3 | NP_766398.1 | 5, 6 | 660 | |||
GenBank mRNA | AK047129.1 | - | 5, 6 | 661 | ||
AY236513.1 | - | 3, 4 | 334 | |||
BC137868.1 | - | 5, 6 | 657 | 8860887 | ||
BC144988.1 | - | 5, 6 | 657 | 9053536 |