Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011842.3 | NP_035972.3 | 14 | 1704 | ||
GenBank mRNA | AF159259.1 | - | 14 | 1579 | ||
AK003701.1 | - | 2 | 138 | |||
AK077548.1 | - | 14 | 1686 | |||
AK088158.1 | - | 14 | 1617 | |||
AK147099.1 | - | 14 | 1683 | |||
AK149834.1 | - | 14 | 1750 | |||
AK157717.1 | - | 14 | 1608 | |||
AK159979.1 | - | 14 | 1680 | |||
AK182140.1 | - | 5 | 397 | |||
AK186547.1 | - | 2 | 138 | |||
AK211323.1 | - | 4 | 327 | |||
AK218441.1 | - | 4 | 327 | |||
AK218740.1 | - | 4 | 327 | |||
BC079847.1 | - | 14 | 1704 | 6419411 |