Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172829.2 | NP_766417.1 | 4 | 1216 | ||
GenBank mRNA | AK082566.1 | - | 4 | 1217 | ||
AK129462.1 | - | 2 | 148 | |||
BC125586.1 | - | 3 | 1086 | 40130101 | ||
BC138142.1 | - | 3 | 1086 | 8861161 |