Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081134.1 | NP_001074603.1 | 1 | 669 | ||
GenBank mRNA | BC147251.1 | - | 1 | 601 | 9056302 | |
BC147252.1 | - | 1 | 601 | 9056297 | ||
BC147715.1 | - | 1 | 601 | 9087474 |