Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182694.2 | NP_874353.2 | 3 | 2583 | ||
NM_182696.2 | NP_874355.1 | 3 | 539 | |||
GenBank mRNA | AF538032.1 | - | 3 | 2072 | ||
AF538033.1 | - | 4 | 1492 | |||
AF538034.1 | - | 3 | 539 | |||
BC089358.1 | - | 3 | 2583 | 30610755 |