Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006080.2 | NP_006071.1 | 10 | 1400 | ||
XM_005250110.3 | XP_005250167.1 | 13 | 1595 | |||
XM_005250111.4 | XP_005250168.1 | 12 | 1435 | |||
XM_006715839.3 | XP_006715902.1 | 11 | 1338 | |||
XM_011515734.2 | XP_011514036.1 | 13 | 1420 | |||
XM_017011673.1 | XP_016867162.1 | 11 | 1255 | |||
GenBank mRNA | AK289954.1 | - | 10 | 1493 | ||
BC111416.1 | - | 10 | 1099 | 40034963 | ||
L26081.1 | - | 10 | 1100 |