Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh11x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271809.1 | NP_001258738.1 | 2 | 455 | ||
NM_001271810.1 | NP_001258739.1 | 2 | 455 | |||
GenBank mRNA | AJ564976.1 | - | 2 | 522 | ||
AK039071.1 | - | 2 | 456 | |||
BC126930.1 | - | 2 | 378 | 40086368 |