Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh11x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271809.1 | NP_001258738.1 | 3 | 2140 | ||
NM_001271810.1 | NP_001258739.1 | 3 | 2140 | |||
GenBank mRNA | AF383875.1 | - | 1 | 559 | ||
AJ564976.1 | - | 3 | 2207 | |||
BC126930.1 | - | 3 | 2063 | 40086368 | ||
EU854474.1 | - | 1 | 902 |