Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081499.2 | NP_001074968.1 | 13 | 2410 | ||
GenBank mRNA | BC051701.1 | - | 376 | |||
BC147581.1 | - | 13 | 2273 | 9056571 | ||
BC151057.1 | - | 13 | 2261 | 9087970 |