Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011830.2 | NP_001011830.2 | 2 | 314 | ||
GenBank mRNA | CB173944.1 | - | 2 | 314 |
Gene Symbol: Olfr121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146629.2 | NP_666840.2 | 2 | 335 | ||
GenBank mRNA | BC090631.1 | - | 2 | 335 | 30358555 |
Gene Symbol: Olfr122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146288.3 | NP_666400.3 | 2 | 311 | ||
GenBank mRNA | CB173933.1 | - | 2 | 311 |