Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197181.1 | NP_001184110.1 | 4 | 646 | ||
NM_006086.3 | NP_006077.2 | 4 | 589 | |||
GenBank mRNA | AF427491.3 | - | 4 | 466 | ||
AK058140.1 | - | 4 | 434 | |||
AK122757.1 | - | 3 | 2548 | |||
AK127418.1 | - | 1928 | ||||
AK223039.1 | - | 4 | 523 | |||
AK292219.1 | - | 4 | 646 | |||
AK314616.1 | - | 4 | 528 | |||
AU142509.1 | - | 4 | 526 | |||
BC000748.2 | - | 4 | 515 | 2823044 | ||
BC001678.2 | - | 3 | 334 | |||
BC003021.1 | - | 4 | 515 | 2823044 | ||
BC047518.1 | - | 2 | 279 | |||
U47634.1 | - | 4 | 466 |