Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146755.2 | NP_666966.1 | 1 | 961 | ||
GenBank mRNA | BC141014.1 | - | 1 | 962 | 9055626 |