Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN2A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197237.1 | NP_001184166.1 | 4 | 783 | ||
NM_001197238.1 | NP_001184167.1 | 4 | 884 | |||
NM_006995.4 | NP_008926.2 | 4 | 761 | |||
NM_181531.2 | NP_853509.1 | 3 | 413 | |||
XM_005248797.3 | XP_005248854.1 | 4 | 778 | |||
XM_005248798.4 | XP_005248855.1 | 3 | 738 | |||
XM_006714953.3 | XP_006715016.1 | 4 | 1030 | |||
XM_006714955.3 | XP_006715018.1 | 4 | 778 | |||
XM_011514228.2 | XP_011512530.1 | 4 | 1083 | |||
XM_011514229.2 | XP_011512531.1 | 4 | 592 | |||
XM_017010166.1 | XP_016865655.1 | 4 | 781 | |||
XM_017010167.1 | XP_016865656.1 | 3 | 683 | |||
XM_017010168.1 | XP_016865657.1 | 4 | 1034 | |||
XM_017010169.1 | XP_016865658.1 | 4 | 752 | |||
GenBank mRNA | AK289883.1 | - | 3 | 401 | ||
AK304744.1 | - | 3 | 412 | |||
BC013258.2 | - | 2 | 333 | |||
BC014021.2 | - | 4 | 759 | 4124138 | ||
BC017497.1 | - | 4 | 776 | 4648935 | ||
U90550.1 | - | 4 | 736 |