Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA407185.1 | - | 2 | 201 | ||
AB221554.1 | - | 2 | 184 | |||
AF149820.1 | - | 2 | 178 | |||
AF177755.1 | - | 2 | 166 | |||
AF177756.1 | - | 2 | 151 | |||
AF177757.1 | - | 2 | 1024 | |||
AK035490.1 | - | 2 | 181 | |||
AK076482.1 | - | 2 | 74 | |||
AK141466.1 | - | 2 | 74 | |||
AK145553.1 | - | 2 | 76 | |||
AK157424.1 | - | 2 | 173 | |||
BC016573.1 | - | 2 | 566 | |||
BC147770.1 | - | 2 | 276 | 9087493 | ||
BC147784.1 | - | 2 | 276 | 9087503 | ||
BY063246.1 | - | 2 | 200 | |||
CJ086997.1 | - | 3 | 222 | |||
CK624100.1 | - | 2 | 62 | |||
DV655694.1 | - | 2 | 144 |