Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136089.2 | NP_001129561.1 | 3 | 332 | ||
NM_011922.3 | NP_036052.2 | 3 | 332 | |||
GenBank mRNA | AJ238978.1 | - | 3 | 232 | ||
AK008772.1 | - | 3 | 255 | |||
AK008808.1 | - | 3 | 256 | |||
AK018597.1 | - | 3 | 289 | |||
AK020288.1 | - | 3 | 257 | |||
AK131880.1 | - | 3 | 252 | |||
AK168715.1 | - | 3 | 236 | |||
BC125319.1 | - | 3 | 204 | 40129834 | ||
BC132207.1 | - | 3 | 204 | 40130484 | ||
BY083459.1 | - | 3 | 332 |