Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170669.2 | NP_733769.1 | 3 | 304 | ||
GenBank mRNA | AK008350.1 | - | 4 | 314 | ||
AK010652.1 | - | 4 | 345 | |||
AK011514.1 | - | 4 | 318 | |||
AK012485.1 | - | 4 | 322 | |||
AK012618.1 | - | 4 | 318 | |||
AK019235.1 | - | 4 | 318 | |||
AK028135.1 | - | 4 | 318 | |||
AK032522.1 | - | 4 | 344 | |||
AK152932.1 | - | 4 | 318 | |||
BC051205.1 | - | 3 | 313 | |||
BC054792.1 | - | 3 | 304 | 6832855 | ||
BC055697.1 | - | 3 | 299 | 6411145 | ||
BC076563.1 | - | 4 | 309 | 6411321 | ||
BC081428.1 | - | 3 | 299 | 6412149 | ||
BC086885.1 | - | 4 | 317 | 6763031 | ||
BC087867.1 | - | 4 | 320 | 6543067 |