Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025581.1 | NP_001020752.1 | 3 | 1284 | ||
GenBank mRNA | AK042937.1 | - | 3 | 704 | ||
AK043934.1 | - | 2 | 968 | |||
AK045425.1 | - | 3 | 1286 | |||
BC116289.1 | - | 3 | 1286 | 40094886 | ||
BC116290.1 | - | 3 | 1285 | 40094891 |