Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207523.2 | NP_997406.1 | 3 | 319 | ||
GenBank mRNA | AF452721.1 | - | 3 | 278 | ||
AK028387.1 | - | 3 | 292 | |||
AK152095.1 | - | 3 | 292 | |||
BC016558.1 | - | 2 | 258 | |||
BC026656.1 | - | 3 | 276 | |||
BC029892.1 | - | 3 | 278 | 4163533 | ||
BC086883.1 | - | 3 | 294 | 6756980 | ||
BC086884.1 | - | 3 | 292 | 30122733 | ||
BC096637.1 | - | 3 | 277 | |||
BC125640.1 | - | 3 | 298 | 40130155 | ||
BC125642.1 | - | 3 | 298 | 40130157 | ||
BY271596.1 | - | 3 | 321 | |||
BY585987.1 | - | 2 | 160 |