Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014258.3 | NP_055073.2 | 35 | 3626 | ||
XM_011528487.2 | XP_011526789.1 | 35 | 3662 | |||
XM_011528488.2 | XP_011526790.1 | 35 | 3614 | |||
XM_011528489.2 | XP_011526791.1 | 35 | 3657 | |||
XM_011528490.2 | XP_011526792.1 | 35 | 3638 | |||
XM_011528493.2 | XP_011526795.1 | 34 | 3577 | |||
XM_017027586.1 | XP_016883075.1 | 34 | 3858 | |||
XM_017027587.1 | XP_016883076.1 | 34 | 3855 | |||
XM_017027588.1 | XP_016883077.1 | 34 | 3859 | |||
XM_017027589.1 | XP_016883078.1 | 33 | 3688 | |||
XM_017027590.1 | XP_016883079.1 | 34 | 3551 | |||
XM_017027591.1 | XP_016883080.1 | 26 | 2957 | |||
GenBank mRNA | BC132864.1 | - | 34 | 3495 | 40146886 | |
BC132870.1 | - | 34 | 3497 | 40146892 | ||
BX648091.1 | - | 35 | 3579 | |||
BX649108.1 | - | 35 | 3604 | |||
Y08982.1 | - | 33 | 3405 |