Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006089.2 | NP_006080.1 | 5 | 445 | ||
NR_033717.1 | 5 | 445 | ||||
XM_006724458.1 | XP_006724521.1 | 5 | 409 | |||
XM_017029219.1 | XP_016884708.1 | 5 | 341 | |||
XM_017029220.1 | XP_016884709.1 | 5 | 337 | |||
XM_017029221.1 | XP_016884710.1 | 5 | 479 | |||
GenBank mRNA | AK303064.1 | - | 4 | 311 | ||
AK315116.1 | - | 5 | 409 | |||
BC051913.1 | - | 5 | 387 | |||
BC064617.1 | - | 5 | 402 | 5575894 | ||
Y18004.1 | - | 5 | 379 |