Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082414.2 | NP_001075883.2 | 8 | 1741 | ||
GenBank mRNA | AF003234.1 | - | 2 | 131 | ||
BC028254.1 | - | 8 | 1289 | |||
BC031117.1 | - | 8 | 852 | 5064197 | ||
D89677.2 | - | 14 | 2227 |