Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178253.5 | NP_839984.1 | 13 | 1149 | ||
GenBank mRNA | AK144621.1 | - | 13 | 1089 | ||
AY238938.1 | - | 13 | 1149 | |||
BC119567.2 | - | 13 | 1127 | 40057793 | ||
BC119568.2 | - | 13 | 1133 | 40057794 |