Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem255b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143671.1 | NP_001137143.1 | 5 | 538 | ||
NM_001143672.1 | NP_001137144.1 | 4 | 475 | |||
GenBank mRNA | AK131817.1 | - | 5 | 538 | ||
BC039763.1 | - | 3 | 302 | |||
BC042836.1 | - | 3 | 291 | |||
BC089620.1 | - | 4 | 450 |