Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem255b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143671.1 | NP_001137143.1 | 9 | 1166 | ||
NM_001143672.1 | NP_001137144.1 | 8 | 1103 | |||
GenBank mRNA | AK131817.1 | - | 9 | 1167 | ||
BC039763.1 | - | 7 | 930 | |||
BC042836.1 | - | 7 | 919 | |||
BC089620.1 | - | 8 | 1078 |