Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcl1b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013774.2 | NP_038802.1 | 4 | 556 | ||
GenBank mRNA | AF195492.1 | - | 4 | 556 | ||
AK139906.1 | - | 4 | 996 | |||
BC145148.1 | - | 4 | 524 | 9053699 | ||
BC145788.1 | - | 4 | 524 | 40131091 |