Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013793.2 | NP_038821.2 | 2 | 186 | ||
GenBank mRNA | AF146571.2 | - | 2 | 186 | ||
AY003920.1 | - | 3 | 268 | |||
BC125466.1 | - | 2 | 238 | 40129981 | ||
BC125494.1 | - | 2 | 238 | 40130009 |
Gene Symbol: Klra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016659.3 | NP_057868.2 | 5 | 762 | ||
GenBank mRNA | AK080158.1 | - | 3 | 344 | ||
AY078436.1 | - | 5 | 762 | |||
M25775.1 | - | 3 | 366 | |||
M25812.1 | - | 3 | 300 |