Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190456.1 | NP_001177385.1 | 11 | 1334 | ||
NM_001190457.1 | NP_001177386.1 | 11 | 1613 | |||
NM_001324014.1 | NP_001310943.1 | 12 | 1545 | |||
NM_001324015.1 | NP_001310944.1 | 11 | 1346 | |||
NM_006091.4 | NP_006082.3 | 11 | 1568 | |||
GenBank mRNA | AB010098.1 | - | 11 | 1301 | ||
AB023142.1 | - | 10 | 1272 | |||
AK124711.1 | - | 6 | 1788 | |||
AK289678.1 | - | 11 | 1613 | |||
BC026335.1 | - | 11 | 1284 | 4792415 |