Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stau2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111272.1 | NP_001104742.1 | 11 | 1417 | ||
NM_025303.3 | NP_079579.2 | 12 | 1548 | |||
GenBank mRNA | AF459099.1 | - | 12 | 1498 | ||
AF459100.1 | - | 11 | 1367 | |||
AK015087.1 | - | 5 | 497 | |||
AK031102.1 | - | 12 | 1475 | |||
AK171076.1 | - | 12 | 1514 | |||
BC010300.1 | - | 9 | 1223 | |||
BC025118.1 | - | 11 | 1372 | 3670067 |